| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000463470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000463470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000463470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049562.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049562.hg.1 |
ENST00000368892.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049562.hg.1 |
ENST00000496809.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049562.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01049562.hg.1 |
TC01006371.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000463470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000463470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000463470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000471423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018489.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
ENST00000354473.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
ENST00000368892.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
ENST00000496809.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
NM_001271087.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
NM_001271088.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
NM_005997.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100243150.hg.1 |
TC0100018489.hg.1 |
Clariom D TC |