Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270641.1 | NP_001257570.1 | 2 - 3 | 403 | 73 | |
| GenBank mRNA | AK131052.1 | - | 2 - 3 | 268 | 73 | |
More Information
Set Membership: |
|

JOSD2
Genomic Map