Assay Details
| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270952.1 | NP_001257881.1 | 3 - 4 | 333 | 145 | |
| NM_006002.4 | NP_005993.1 | 3 - 4 | 255 | 145 | ||
| XM_011535212.1 | XP_011533514.1 | 3 - 4 | 255 | 145 | ||
| XM_011535213.2 | XP_011533515.1 | 2 - 3 | 236 | 145 | ||
| XM_011535214.2 | XP_011533516.1 | 3 - 4 | 323 | 145 | ||
| XM_017020725.1 | XP_016876214.1 | 3 - 4 | 255 | 145 | ||
| XM_017020726.1 | XP_016876215.1 | 3 - 4 | 255 | 145 | ||
| XM_017020727.1 | XP_016876216.1 | 2 - 3 | 230 | 145 | ||
| GenBank mRNA | AK313665.1 | - | 3 - 4 | 245 | 145 | |
| BC018125.2 | - | 3 - 4 | 214 | 3887201 | 145 | |
| BQ427804.1 | - | 3 - 4 | 269 | 145 | ||
| BT019359.1 | - | 3 - 4 | 184 | 145 | ||
| CR456855.1 | - | 3 - 4 | 184 | 145 | ||
| DA820150.1 | - | 3 - 4 | 255 | 145 | ||
| M30496.1 | - | 3 - 4 | 204 | 145 | ||
More Information
Set Membership: |
|

UCHL3
Genomic Map