Assay Details
| Gene Symbol: UCHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270952.1 | NP_001257881.1 | 6 - 7 | 629 | 87 | |
| NM_006002.4 | NP_005993.1 | 6 - 7 | 551 | 87 | ||
| XM_017020727.1 | XP_016876216.1 | 5 - 6 | 526 | 87 | ||
| GenBank mRNA | AK313665.1 | - | 6 - 7 | 541 | 87 | |
| BC018125.2 | - | 6 - 7 | 510 | 3887201 | 87 | |
| BQ427804.1 | - | 6 - 7 | 565 | 87 | ||
| BT019359.1 | - | 6 - 7 | 480 | 87 | ||
| CR456855.1 | - | 6 - 7 | 480 | 87 | ||
| M30496.1 | - | 6 - 7 | 500 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300166720.hg.1 | TC1300009979.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000377595.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000419068.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000471792.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000606347.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000607339.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | ENST00000618773.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_001270952.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009979.hg.1 | NM_006002.4 | Refseq |

UCHL3
Genomic Map