Assay Details
| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270891.1 | NP_001257820.1 | 2 - 3 | 176 | 112 | |
| NM_024108.2 | NP_077013.1 | 2 - 3 | 218 | 112 | ||
| XM_017027295.1 | XP_016882784.1 | 2 - 3 | 277 | 112 | ||
| GenBank mRNA | AF161407.1 | - | 2 - 3 | 170 | 112 | |
| BC001907.1 | - | 2 - 3 | 202 | 3534111 | 112 | |
| BC004450.1 | - | 2 - 3 | 203 | 2820830 | 112 | |
| CN256687.1 | - | 2 - 3 | 188 | 112 | ||
| FJ418644.1 | - | 2 - 3 | 158 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001615.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19022040.hg.1 | ENST00000006275.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022040.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19022040.hg.1 | TC19001615.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010926.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_024108.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | TC1900010926.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000006275.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000585934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000587818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000588062.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | ENST00000592647.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_001270891.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_001270892.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_001270893.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900190658.hg.1 | NM_024108.2 | Refseq |

TRAPPC6A
Genomic Map