Assay Details
| Gene Symbol: SKOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258024.1 | NP_001244953.1 | 6 - 7 | 1588 | 90 | |
| GenBank mRNA | AY669507.1 | - | 6 - 7 | 1588 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
transcription corepressor activity
SMAD binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
transcription corepressor activity
SMAD binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000560577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000560577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | ENST00000560577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500154337.hg.1 | TC1500007652.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000341418.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000380035.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554054.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000554240.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000558889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000560577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000560577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | ENST00000560577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007652.hg.1 | NM_001258024.1 | Refseq |

SKOR1
Genomic Map