Assay Details
| Gene Symbol: GAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012296.3 | NP_036428.1 | 10 | 4179 | 86 | |
| NM_080491.2 | NP_536739.1 | 10 | 4235 | 86 | ||
| XM_006718753.2 | XP_006718816.1 | 11 | 4280 | 86 | ||
| XM_011545408.2 | XP_011543710.1 | 9 | 3886 | 86 | ||
| GenBank mRNA | AB011143.1 | - | 10 | 4179 | 86 | |
| AB018413.1 | - | 10 | 4133 | 86 | ||
| BC152459.1 | - | 10 | 4179 | 8862595 | 86 | |
More Information
Set Membership: |
|

GAB2
Genomic Map