Assay Details
| Gene Symbol: GSKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271906.1 | NP_001258835.1 | 1 - 2 | 119 | 94 | |
| GenBank mRNA | HY108584.1 | - | 1 - 2 | 95 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271906.1 transcripts.
Set Membership: |
|

GSKIP
Genomic Map