Assay Details
| Gene Symbol: VSIG10L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163922.1 | NP_001157394.1 | 1 - 2 | 37 | 87 | |
| GenBank mRNA | BC131493.1 | - | 1 - 2 | 37 | 87 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000594311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000594311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000594311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000335624.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | ENST00000600663.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011248.hg.1 | NM_001163922.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000594311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000594311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000594311.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008639.hg.1 | ENST00000601148.1 | Ensembl |

VSIG10L
Genomic Map