Assay Details
| Gene Symbol: PTS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000317.2 | NP_000308.1 | 5 - 6 | 388 | 97 | |
| GenBank mRNA | BC009686.1 | - | 5 - 6 | 352 | 3901665 | 97 |
| D17400.1 | - | 5 - 6 | 377 | 97 | ||
| M97655.1 | - | 5 - 6 | 318 | 97 | ||
More Information
Set Membership: |
|

PTS
Genomic Map