Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4736 | MIR4736 | -1.2kb | Anti-sense | Chr. 17 - 58335976 - 58336022 on NCBI Build GRCh38 | |
| Downstream | heat shock transcription factor 5 | HSF5 |
HSF 5, HSTF 5 |
25.2kb | Anti-sense | Chr. 17 - 58420167 - 58488401 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map