| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000424788.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000426549.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000439077.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008297.hg.1 |
ENST00000597638.1 |
Ensembl |