Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7702 | MIR7702 | hsa-mir-7702 | -1.6kb | Sense | Chr. 9 - 111271156 - 111271214 on NCBI Build GRCh38 |
| Downstream | olfactory receptor family 2 subfamily K member 2 | OR2K2 |
HSHTPCRH06, HTPCRH06, OR2AN1P, OR2AR1P |
42.5kb | Sense | Chr. 9 - 111326884 - 111330224 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map