Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105372969 | LOC105372969 | Sense | Chr. 22 - 25280703 - 25283094 on NCBI Build GRCh38 | ||
| Downstream |
immunoglobulin lambda like polypeptide 3, pseudogene |
IGLL3P |
16.1, IGLL3 |
36.0kb | Anti-sense | Chr. 22 - 25318257 - 25320227 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map