Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead associated phosphopeptide binding domain 1 | FHAD1 | Anti-sense | Chr. 1 - 15236559 - 15400283 on NCBI Build GRCh38 | ||
| Downstream | uncharacterized LOC101927441 | LOC101927441 | 68.3kb | Sense | Chr. 1 - 15402979 - 15413823 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map