Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105369819 | LOC105369819 | -7.1kb | Sense | Chr. 12 - 68332519 - 68337612 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100507195 | LOC100507195 | 81.9kb | Anti-sense | Chr. 12 - 68431842 - 68451484 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map