Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724663 | LOC102724663 | -104.9kb | Sense | Chr. 12 - 81279189 - 81312422 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928449 | LOC101928449 | 523.4kb | Sense | Chr. 12 - 81953719 - 81993133 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map