Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105371397 | LOC105371397 | -37.6kb | Anti-sense | Chr. 16 - 87700832 - 87703017 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC102724467 | LOC102724467 | 18.6kb | Anti-sense | Chr. 16 - 87766331 - 87779148 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map