Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MFI2 antisense RNA 1 | MFI2-AS1 | Anti-sense | Chr. 3 - 197002906 - 197004744 on NCBI Build GRCh38 | ||
| Downstream | discs large MAGUK scaffold protein 1 | DLG1 |
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg |
37.8kb | Sense | Chr. 3 - 197042560 - 197299272 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map