Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S2 pseudogene 47 | RPS2P47 | RPS2_25_1553 | -1.7kb | Anti-sense | Chr. 17 - 47159154 - 47159998 on NCBI Build GRCh38 |
| Downstream | myosin light chain 4 | MYL4 |
ALC1, AMLC, GT1, PRO1957 |
46.9kb | Sense | Chr. 17 - 47209017 - 47227650 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map