Assay Details
| Gene Symbol: TMPRSS2-ERG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMPRSS2-ERG_T1E7.0 | - | 93 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS2-ERG fusion| Gene Symbol: TMPRSS2-ERG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMPRSS2-ERG_T1E7.0 | - | 93 | 88 | ||