Assay Details
| Gene Symbol: TMPRSS2-ETV1a fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMPRSS2-ETV1a_T1E4.0 | - | 47 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS2-ETV1a fusion| Gene Symbol: TMPRSS2-ETV1a fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMPRSS2-ETV1a_T1E4.0 | - | 47 | 115 | ||
Set Membership: |
|