Assay Details
| Gene Symbol: WIPF2-ERBB2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_WIPF2-ERBB2_W1E4.0 | - | 69 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WIPF2-ERBB2 fusion| Gene Symbol: WIPF2-ERBB2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_WIPF2-ERBB2_W1E4.0 | - | 69 | 97 | ||