Assay Details
| Gene Symbol: SND1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JX013981.1 | - | 2030 | 89 | ||
| OM_SND1-BRAF_S16B9.0 | - | 92 | 89 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SND1-BRAF fusion| Gene Symbol: SND1-BRAF fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JX013981.1 | - | 2030 | 89 | ||
| OM_SND1-BRAF_S16B9.0 | - | 92 | 89 | |||