Assay Details
| Gene Symbol: TMPRSS2-ETV4a fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_TMPRSS2-ETV4a_T1bE3.0 | - | 71 | 74 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

TMPRSS2-ETV4a fusion
Genomic Map