Assay Details
| Gene Symbol: FGFR2-KIAA1967 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-KIAA1967_CCAR2_F17C4.0 | - | 92 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FGFR2-KIAA1967 fusion| Gene Symbol: FGFR2-KIAA1967 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_FGFR2-KIAA1967_CCAR2_F17C4.0 | - | 92 | 87 | ||