Assay Details
| Gene Symbol: ESRP1-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | GU149302.1 | - | 1983 | 90 | ||
| OM_ESRP1-RAF1_E13R6_COSF826.0 | - | 47 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESRP1-RAF1 fusion| Gene Symbol: ESRP1-RAF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | GU149302.1 | - | 1983 | 90 | ||
| OM_ESRP1-RAF1_E13R6_COSF826.0 | - | 47 | 90 | |||