Assay Details
| Gene Symbol: HOOK3-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ104207.1 | - | 1093 | 99 | ||
| OM_HOOK3-RET_H11R12.0 | - | 49 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOOK3-RET fusion| Gene Symbol: HOOK3-RET fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ104207.1 | - | 1093 | 99 | ||
| OM_HOOK3-RET_H11R12.0 | - | 49 | 99 | |||