Assay Details
| Gene Symbol: PAX8-PPARG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PAX8-PPARG_P10P2_COSF1219.0 | - | 23 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAX8-PPARG fusion| Gene Symbol: PAX8-PPARG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_PAX8-PPARG_P10P2_COSF1219.0 | - | 23 | 71 | ||