Assay Details
| Gene Symbol: SLC45A3-ERG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SLC45A3-ERG_S1E4_COSF1138.0 | - | 50 | 86 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC45A3-ERG fusion| Gene Symbol: SLC45A3-ERG fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_SLC45A3-ERG_S1E4_COSF1138.0 | - | 50 | 86 | ||