Assay Details
| Gene Symbol: STRN-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_STRN-ALK_S3A20_COSF1430.0 | - | 98 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STRN-ALK fusion| Gene Symbol: STRN-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | OM_STRN-ALK_S3A20_COSF1430.0 | - | 98 | 94 | ||