Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000428559.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000526521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000526530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000528821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000530274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000532573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000532964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000533447.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
NM_001300779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
NM_015954.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000428559.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000526521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000526530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000528821.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000530274.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000532573.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000532964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
ENST00000533447.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
NM_001300779.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
NM_015954.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200148725.hg.1 |
TC1200006967.hg.1 |
Clariom D TC |