Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300783.1 | NP_001287712.1 | 1 - 2 | 367 | 131 | |
| GenBank mRNA | BC092474.1 | - | 1 - 2 | 368 | 30528467 | 131 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157963.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157963.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157963.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157963.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157963.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157963.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157963.hg.1 | TC0500008454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157965.hg.1 | TC0500008454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008454.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157964.hg.1 | TC0500008454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000379551.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000407149.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000446965.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000505123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | ENST00000509923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_001300783.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_001308087.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_001308088.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | NM_016644.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500157975.hg.1 | TC0500008454.hg.1 | Clariom D TC |

PRR16
Genomic Map