Assay Details
| Gene Symbol: CIRBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280.2 | NP_001271.1 | 4 - 5 | 502 | 140 | |
| NM_001300815.1 | NP_001287744.1 | 4 - 5 | 504 | 140 | ||
| NM_001300829.1 | NP_001287758.1 | 4 - 5 | 504 | 140 | ||
| NR_023312.2 | 4 - 5 | 563 | 140 | |||
| NR_023313.2 | 3 - 4 | 395 | 140 | |||
| XM_006722637.1 | XP_006722700.1 | 4 - 5 | 565 | 140 | ||
| XM_011527668.1 | XP_011525970.1 | 4 - 5 | 438 | 140 | ||
| XM_017026237.1 | XP_016881726.1 | 4 - 5 | 565 | 140 | ||
| GenBank mRNA | AB451225.1 | - | 3 - 4 | 353 | 140 | |
| AB451348.1 | - | 3 - 4 | 353 | 140 | ||
| AF021336.1 | - | 4 - 5 | 435 | 140 | ||
| AK094781.1 | - | 3 - 4 | 328 | 140 | ||
| AK095781.1 | - | 2 - 3 | 623 | 140 | ||
| AK304185.1 | - | 3 - 4 | 581 | 140 | ||
| AK311601.1 | - | 4 - 5 | 457 | 140 | ||
| BC000403.2 | - | 4 - 5 | 414 | 2822968 | 140 | |
| BC000901.1 | - | 4 - 5 | 419 | 2900585 | 140 | |
| BM706055.1 | - | 4 - 5 | 416 | 140 | ||
| BP195090.1 | - | 4 - 5 | 502 | 140 | ||
| BT007210.1 | - | 3 - 4 | 353 | 140 | ||
| D78134.1 | - | 4 - 5 | 433 | 140 | ||
| DA433715.1 | - | 4 - 5 | 439 | 140 | ||
More Information
Set Membership: |
|

CIRBP
Genomic Map