Assay Details
| Gene Symbol: FOSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300844.1 | NP_001287773.1 | 1 - 2 | 289 | 79 | |
| NM_001300855.1 | NP_001287784.1 | 1 - 2 | 289 | 79 | ||
| NM_005438.4 | NP_005429.1 | 1 - 2 | 289 | 79 | ||
| GenBank mRNA | AK222902.1 | - | 1 - 2 | 146 | 79 | |
| AK293297.1 | - | 1 - 2 | 151 | 79 | ||
| BC016648.1 | - | 1 - 2 | 147 | 3925241 | 79 | |
| BM764259.1 | - | 1 - 2 | 182 | 79 | ||
| BP222547.1 | - | 1 - 2 | 182 | 79 | ||
| BQ937004.1 | - | 1 - 2 | 289 | 79 | ||
| CR542257.1 | - | 1 - 2 | 102 | 79 | ||
| CR542278.1 | - | 1 - 2 | 102 | 79 | ||
| KU178407.1 | - | 1 - 2 | 102 | 79 | ||
| KU178408.1 | - | 1 - 2 | 102 | 79 | ||
| X16707.1 | - | 1 - 2 | 136 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FOSL1
Genomic Map