Assay Details
| Gene Symbol: OAZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134939.1 | NP_001128411.1 | 2 - 3 | 339 | 69 | |
| NM_001301371.1 | NP_001288300.1 | 2 - 3 | 226 | 69 | ||
| NM_016178.2 | NP_057262.2 | 2 - 3 | 374 | 69 | ||
| GenBank mRNA | AF175296.1 | - | 2 - 3 | 374 | 69 | |
| AY713300.1 | - | 2 - 3 | 313 | 69 | ||
| BC073949.1 | - | 2 - 3 | 338 | 5416827 | 69 | |
| BI830988.1 | - | 2 - 3 | 226 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cellular amino acid metabolic process
polyamine biosynthetic process
spermatogenesis
putrescine transport
negative regulation of catalytic activity
positive regulation of protein catabolic process
negative regulation of polyamine transmembrane transport
protein binding
ornithine decarboxylase inhibitor activity
putrescine transmembrane transporter activity
regulation of cellular amino acid metabolic process
polyamine biosynthetic process
spermatogenesis
putrescine transport
negative regulation of catalytic activity
positive regulation of protein catabolic process
negative regulation of polyamine transmembrane transport
protein binding
ornithine decarboxylase inhibitor activity
putrescine transmembrane transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000420382.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000479764.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000582415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009974.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000321531.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000400999.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000453029.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000577465.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | ENST00000627780.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | NM_001134939.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | NM_001301371.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | NM_016178.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100172327.hg.1 | TC0100009974.hg.1 | Clariom D TC |

OAZ3
Genomic Map