Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124.2 | NP_001115.1 | 2 - 3 | 368 | 88 | |
| GenBank mRNA | AK312893.1 | - | 2 - 3 | 275 | 88 | |
| BC015961.2 | - | 2 - 3 | 263 | 3506276 | 88 | |
| BT006902.1 | - | 1 - 2 | 97 | 88 | ||
| CR541995.1 | - | 1 - 2 | 97 | 88 | ||
| D14874.1 | - | 2 - 3 | 253 | 88 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
response to hypoxia
neural tube closure
response to yeast
regulation of the force of heart contraction
G-protein coupled receptor internalization
cAMP biosynthetic process
progesterone biosynthetic process
signal transduction
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
heart development
female pregnancy
aging
blood circulation
androgen metabolic process
positive regulation of cell proliferation
negative regulation of cell proliferation
response to cold
response to wounding
positive regulation of heart rate
antibacterial humoral response
antifungal humoral response
cAMP-mediated signaling
positive regulation of cAMP biosynthetic process
organ regeneration
neuron projection regeneration
receptor internalization
response to lipopolysaccharide
response to insulin
odontogenesis of dentin-containing tooth
response to starvation
positive regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of angiogenesis
negative regulation of vasoconstriction
positive regulation of vasodilation
hormone secretion
developmental growth
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
response to glucocorticoid
branching involved in labyrinthine layer morphogenesis
spongiotrophoblast layer development
vascular smooth muscle cell development
positive regulation of vasculogenesis
receptor binding
hormone activity
protein binding
adrenomedullin receptor binding
vasculogenesis
response to hypoxia
neural tube closure
response to yeast
regulation of the force of heart contraction
G-protein coupled receptor internalization
cAMP biosynthetic process
progesterone biosynthetic process
signal transduction
positive regulation of cytosolic calcium ion concentration
cell-cell signaling
heart development
female pregnancy
aging
blood circulation
androgen metabolic process
positive regulation of cell proliferation
negative regulation of cell proliferation
response to cold
response to wounding
positive regulation of heart rate
antibacterial humoral response
antifungal humoral response
cAMP-mediated signaling
positive regulation of cAMP biosynthetic process
organ regeneration
neuron projection regeneration
receptor internalization
response to lipopolysaccharide
response to insulin
odontogenesis of dentin-containing tooth
response to starvation
positive regulation of apoptotic process
negative regulation of vascular permeability
positive regulation of angiogenesis
negative regulation of vasoconstriction
positive regulation of vasodilation
hormone secretion
developmental growth
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
response to glucocorticoid
branching involved in labyrinthine layer morphogenesis
spongiotrophoblast layer development
vascular smooth muscle cell development
positive regulation of vasculogenesis
receptor binding
hormone activity
protein binding
adrenomedullin receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528544.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000278175.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000524948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000525063.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000526492.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528544.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000528655.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000530439.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | ENST00000534464.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006831.hg.1 | NM_001124.2 | Refseq |

ADM
Genomic Map