| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007015.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007015.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007015.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007015.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007015.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147716.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147716.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147716.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147716.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147716.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147716.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147716.hg.1 |
TC0X00007015.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00147717.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147717.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147717.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147717.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147717.hg.1 |
ENST00000456333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147717.hg.1 |
NM_001289773.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147717.hg.1 |
TC0X00007015.hg.1 |
Clariom D TC |