Assay Details
| Gene Symbol: HSPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130106.1 | NP_001123578.1 | 4 - 5 | 747 | 59 | |
| NM_001297600.1 | NP_001284529.1 | 2 - 3 | 662 | 59 | ||
| NM_012267.4 | NP_036399.3 | 3 - 4 | 856 | 59 | ||
| XM_005258700.2 | XP_005258757.1 | 3 - 4 | 587 | 59 | ||
| XM_005258701.2 | XP_005258758.1 | 3 - 4 | 649 | 59 | ||
| XM_011526691.1 | XP_011524993.1 | 4 - 5 | 745 | 59 | ||
| XM_017026543.1 | XP_016882032.1 | 3 - 4 | 584 | 59 | ||
| XM_017026544.1 | XP_016882033.1 | 3 - 4 | 584 | 59 | ||
| XM_017026545.1 | XP_016882034.1 | 3 - 4 | 587 | 59 | ||
| GenBank mRNA | AB020592.1 | - | 3 - 4 | 578 | 59 | |
| AF093420.1 | - | 3 - 4 | 581 | 59 | ||
| AF187859.1 | - | 3 - 4 | 546 | 59 | ||
| AF217996.1 | - | 4 - 5 | 733 | 59 | ||
| AK075293.1 | - | 3 - 4 | 603 | 59 | ||
| AK129755.1 | - | 3 - 4 | 659 | 59 | ||
| AK130636.1 | - | 609 | 59 | |||
| AK130781.1 | - | 3 - 4 | 593 | 59 | ||
| AK294358.1 | - | 2 - 3 | 661 | 59 | ||
| BC001236.1 | - | 2 - 3 | 511 | 3454675 | 59 | |
| BC002373.2 | - | 3 - 4 | 568 | 2822836 | 59 | |
| CR457118.1 | - | 2 - 3 | 413 | 59 | ||
More Information
Set Membership: |
|

HSPBP1
Genomic Map