Assay Details
| Gene Symbol: ART5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297668.1 | NP_001284597.1 | 3 - 4 | 1069 | 80 | |
| XM_005252779.3 | XP_005252836.1 | 3 - 4 | 949 | 284 | ||
| GenBank mRNA | AY358466.1 | - | 3 - 4 | 694 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000359918.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397067.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000397068.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000425767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000425767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000425767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000425767.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000453353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000453353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000453353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000453353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000453353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000453353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | ENST00000453353.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001079536.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_001297668.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NM_053017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NR_123732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NR_123732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NR_123732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NR_123732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NR_123732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NR_123732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009855.hg.1 | NR_123732.1 | Refseq |

ART5
Genomic Map