Assay Details
| Gene Symbol: LETMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243689.1 | NP_001230618.1 | 2 - 3 | 331 | 130 | |
| NM_001300765.1 | NP_001287694.1 | 2 - 3 | 331 | 130 | ||
| NM_015416.4 | NP_056231.3 | 2 - 3 | 331 | 130 | ||
| NR_045017.1 | 2 - 3 | 206 | 130 | |||
| XM_006719339.1 | XP_006719402.1 | 2 - 3 | 303 | 130 | ||
| XM_011538161.2 | XP_011536463.1 | 2 - 3 | 338 | 130 | ||
| XM_011538162.2 | XP_011536464.1 | 2 - 3 | 339 | 130 | ||
| XM_011538163.2 | XP_011536465.1 | 2 - 3 | 340 | 130 | ||
| XM_011538165.2 | XP_011536467.1 | 2 - 3 | 338 | 130 | ||
| XM_017019151.1 | XP_016874640.1 | 2 - 3 | 340 | 130 | ||
| XM_017019152.1 | XP_016874641.1 | 2 - 3 | 306 | 130 | ||
| XM_017019155.1 | XP_016874644.1 | 2 - 3 | 178 | 130 | ||
| XM_017019159.1 | XP_016874648.1 | 2 - 3 | 341 | 130 | ||
| XM_017019160.1 | XP_016874649.1 | 2 - 3 | 303 | 130 | ||
| XM_017019165.1 | XP_016874654.1 | 2 - 3 | 303 | 130 | ||
| XM_017019167.1 | XP_016874656.1 | 2 - 3 | 303 | 130 | ||
| XM_017019168.1 | XP_016874657.1 | 2 - 3 | 303 | 130 | ||
| GenBank mRNA | AF195651.2 | - | 2 - 3 | 303 | 130 | |
| AF315598.1 | - | 2 - 3 | 167 | 130 | ||
| AK000639.1 | - | 2 - 3 | 318 | 130 | ||
| AK057866.1 | - | 2 - 3 | 276 | 130 | ||
| AK127540.1 | - | 2 - 3 | 292 | 130 | ||
| AK225836.1 | - | 2 - 3 | 289 | 130 | ||
| AK301955.1 | - | 2 - 3 | 206 | 130 | ||
| AK315119.1 | - | 2 - 3 | 130 | |||
| BC064943.1 | - | 2 - 3 | 277 | 6009854 | 130 | |
| BX376670.2 | - | 2 - 3 | 272 | 130 | ||
| DC357852.1 | - | 2 - 3 | 331 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006719339.1, XM_017019165.1 transcripts.
Set Membership: |
|

LETMD1
Genomic Map