Assay Details
| Gene Symbol: LYPLAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300770.1 | NP_001287699.1 | 3 - 4 | 374 | 154 | |
| NM_001300771.1 | NP_001287700.1 | 3 - 4 | 405 | 154 | ||
| NM_001300772.1 | NP_001287701.1 | 3 - 4 | 323 | 154 | ||
| NM_138794.4 | NP_620149.2 | 3 - 4 | 422 | 154 | ||
| XM_006711162.3 | XP_006711225.1 | 4 - 5 | 401 | 154 | ||
| XM_011509182.2 | XP_011507484.1 | 4 - 5 | 433 | 154 | ||
| XM_011509183.2 | XP_011507485.1 | 4 - 5 | 734 | 154 | ||
| XM_017000271.1 | XP_016855760.1 | 4 - 5 | 433 | 154 | ||
| XM_017000272.1 | XP_016855761.1 | 3 - 4 | 638 | 154 | ||
| GenBank mRNA | AA417842.1 | - | 3 - 4 | 321 | 154 | |
| AK291542.1 | - | 3 - 4 | 376 | 154 | ||
| AY341430.1 | - | 3 - 4 | 314 | 154 | ||
| BC016711.1 | - | 3 - 4 | 376 | 4133970 | 154 | |
| BX282451.1 | - | 3 - 4 | 321 | 154 | ||
| CB851909.1 | - | 3 - 4 | 412 | 154 | ||
| DA822882.1 | - | 3 - 4 | 403 | 154 | ||
More Information
Set Membership: |
|

LYPLAL1
Genomic Map