Assay Details
| Gene Symbol: PQLC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282710.1 | NP_001269639.1 | 1 - 2 | 315 | 63 | |
| NM_001282711.1 | NP_001269640.1 | 1 - 2 | 315 | 63 | ||
| NM_001282712.1 | NP_001269641.1 | 1 - 2 | 315 | 63 | ||
| NM_152391.4 | NP_689604.1 | 1 - 2 | 315 | 63 | ||
| NR_104231.1 | 1 - 2 | 315 | 63 | |||
| XM_011510315.1 | XP_011508617.1 | 1 - 2 | 315 | 63 | ||
| GenBank mRNA | AK001091.2 | - | 1 - 2 | 231 | 63 | |
| AK301439.1 | - | 1 - 2 | 210 | 63 | ||
| AK313402.1 | - | 1 - 2 | 210 | 63 | ||
| AL548337.3 | - | 1 - 2 | 86 | 63 | ||
| BC027625.1 | - | 1 - 2 | 279 | 4825091 | 63 | |
| BI601351.1 | - | 1 - 2 | 261 | 63 | ||
| BQ277359.1 | - | 1 - 2 | 79 | 63 | ||
| CX751769.1 | - | 1 - 2 | 152 | 63 | ||
| DB460796.1 | - | 1 - 2 | 315 | 63 | ||
| DC392668.1 | - | 1 - 2 | 299 | 63 | ||
More Information
Set Membership: |
|

PQLC3
Genomic Map