Assay Details
| Gene Symbol: TMEM139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104251.1 | 1 - 2 | 74 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105375545 | LOC105375545 | Anti-sense | Chr. 7 - 143255299 - 143286334 on NCBI Build GRCh38 | ||
| Downstream | caspase 2 | CASP2 |
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57 |
166.0 | Sense | Chr. 7 - 143288215 - 143307696 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167380.hg.1 | TC0700009487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000480421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000487419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000480421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | ENST00000487419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167398.hg.1 | TC0700009487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000480421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000487419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000480421.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | ENST00000487419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167397.hg.1 | TC0700009487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000487419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | ENST00000487419.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167399.hg.1 | TC0700009487.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000359333.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000409102.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000409244.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000409541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000410004.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | ENST00000471161.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001242773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001242774.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001242775.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001282876.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_001282877.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NM_153345.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_040003.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104250.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104251.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104252.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104253.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | NR_104254.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700167400.hg.1 | TC0700009487.hg.1 | Clariom D TC |

TMEM139
Genomic Map