Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000298068.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000371667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000446394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000498446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
NM_001284274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
NM_182765.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
NR_104291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000298068.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000371667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000446394.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
ENST00000498446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
NM_001284274.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
NM_182765.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
NR_104291.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000157299.hg.1 |
TC1000008435.hg.1 |
Clariom D TC |