Assay Details
| Gene Symbol: HACL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104315.1 | 4 - 5 | 697 | 71 | ||
| GenBank mRNA | BC007440.2 | - | 4 - 5 | 402 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | collagen like tail subunit of asymmetric acetylcholinesterase | COLQ |
CMS5, EAD |
-64.8kb | Sense | Chr. 3 - 15450133 - 15521751 on NCBI Build GRCh38 |
| Downstream | biotinidase | BTD | 9.7kb | Anti-sense | Chr. 3 - 15601352 - 15653709 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

HACL1
Genomic Map