| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174854.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174854.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174854.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174854.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174854.hg.1 |
TC0200010569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200174857.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174857.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174857.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174857.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174857.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174857.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174857.hg.1 |
TC0200010569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
ENST00000624973.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010569.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174855.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174855.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174855.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174855.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174855.hg.1 |
TC0200010569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200174851.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174851.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174851.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174851.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174851.hg.1 |
TC0200010569.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200174852.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174852.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174852.hg.1 |
ENST00000623040.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200174852.hg.1 |
NR_104359.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200174852.hg.1 |
TC0200010569.hg.1 |
Clariom D TC |