| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000416598.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000420811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000432897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000432897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000434141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000457412.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000457412.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000587466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000587466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000590080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000590080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591523.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591523.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000608806.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000608833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000610175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000610197.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104362.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104364.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000416598.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000420811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000432897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000432897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000434141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000457412.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000457412.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585576.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000585589.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000587466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000587466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588058.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000588227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000590080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000590080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591173.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591523.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591523.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000591882.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592838.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000592898.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000608806.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000608833.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000610175.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
ENST00000610197.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104362.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104363.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104364.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
NR_104365.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100160174.hg.1 |
TC0100008467.hg.1 |
Clariom D TC |