Assay Details
| Gene Symbol: CLPSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104469.1 | 1 - 2 | 211 | 104 | ||
| GenBank mRNA | AA861820.1 | - | 1 - 2 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | armadillo repeat containing 12 | ARMC12 | C6orf81 | -27.7kb | Sense | Chr. 6 - 35733726 - 35748913 on NCBI Build GRCh38 |
| Downstream | colipase like 1 | CLPSL1 |
C6orf127, ESP32, dJ510O8.6 |
1.5kb | Sense | Chr. 6 - 35781054 - 35794039 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007819.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154142.hg.1 | TC0600007819.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000360454.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000403376.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000467122.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | ENST00000481904.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NM_001286550.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NM_207409.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NR_104467.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | NR_104469.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600154141.hg.1 | TC0600007819.hg.1 | Clariom D TC |

CLPSL2
Genomic Map