Assay Details
| Gene Symbol: LOC101928505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104668.1 | 2 - 3 | 365 | 131 | ||
| GenBank mRNA | BX101031.1 | - | 2 - 3 | 413 | 131 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LincR-0003 | LINCR-0003 | -47.8kb | Sense | Chr. 5 - 57570342 - 57617430 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928539 | LOC101928539 | 219.3kb | Anti-sense | Chr. 5 - 57890331 - 57899162 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928505
Genomic Map